Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIR-NKAT2 Mouse anti-Human, BV480, Clone: DX27, BD OptiBuild
Mouse Monoclonal Antibody
Manufacturer: BD Biosciences 746573
Description
KIR-NKAT2 Monoclonal antibody specifically detects KIR-NKAT2 in Human samples. It is validated for Flow Cytometry.Specifications
KIR-NKAT2 | |
DX27 | |
Brilliant Violet 480 | |
Mouse | |
50 μg | |
Refrigerate (2 to 8°C) | |
Monoclonal |
Flow Cytometry | |
0.2 mg/mL | |
Purified | |
IgG2a, κ | |
RUO | |
Primary | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title