Learn More
Kir2.2 (KCNJ12) Mouse anti-Human, Mouse, Rat, Clone: S124B-38, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA527678
Catalog No. PIMA527678
Add to cart
Description
Description
Inward rectifying potassium channel that is activated by phosphatidylinositol 4,5-bisphosphate and that probably participates in controlling the resting membrane potential in electrically excitable cells. Probably participates in establishing action potential waveform and excitability of neuronal and muscle tissues. Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium.
Specifications
Specifications
Kir2.2 (KCNJ12)
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
P52187, P52188, Q14500
IRK2, KCNJ12, kcnj12x, KCNJN1, Kir2.2v, potassium inwardly rectifying channel subfamily J member 12
Mouse
IgG1
100 μg
-20°C
Primary
117052, 16515, 3768
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
S124B-38
Unconjugated
KCNJ12
Liquid
KCNJ12
Fusion protein amino acids 362-427 of rat Kir2.2
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat