Learn More
Kir2.3 (KCNJ4) Mouse anti-Human, Mouse, Rat, Clone: S25-35, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA527680
Catalog No. PIMA527680
Add to cart
Description
Description
Several different potassium channels are known to be involved with electrical signaling in the nervous system. One class is activated by depolarization whereas a second class is not. The latter are referred to as inwardly rectifying K+ channels, and they have a greater tendency to allow potassium to flow into the cell rather than out of it. This asymmetry in potassium ion conductance plays a key role in the excitability of muscle cells and neurons. The protein encoded by this gene is an integral membrane protein and member of the inward rectifier potassium channel family. The encoded protein has a small unitary conductance compared to other members of this protein family. Two transcript variants encoding the same protein have been found for this gene.
Specifications
Specifications
Kir2.3 (KCNJ4)
Monoclonal
1 mg/ml
PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
P48050, P52189, P52190
IRK3, HIR, HIRK2, HRK1, KCNJ4, potassium inwardly rectifying channel subfamily J member 4
Mouse
IgG1
100 μg
-20°C
Primary
116649, 16520, 3761
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
S25-35
Unconjugated
KCNJ4
Liquid
Kcnj4
Fusion protein amino acids 390-445 of human Kir2.3
Protein G
RUO
Antibody
Monoclonal
Human, Mouse, Rat