Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KIR2DL1/CD158a Mouse anti-Human, Alexa Fluor™ 750, Clone: MM0439-8J29, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211759AF750
Description
KIR2DL1/CD158a Monoclonal specifically detects KIR2DL1/CD158a in Human samples. It is validated for Flow Cytometry.Specifications
KIR2DL1/CD158a | |
Monoclonal | |
Alexa Fluor 750 | |
50 mM sodium borate with 0.05% sodium azide | |
KIR2DL1 | |
Human recombinant KIR2DL1/CD158a | |
0.1 mL | |
Primary | |
This antibody only detects KIR2DL1 there is no cross reactivity with other KIRs. | |
Store at 4°C in the dark. |
Flow Cytometry | |
MM0439-8J29 | |
Flow Cytometry | |
CD158 antigen-like family member A, CD158a antigen, CD158Ankat1, cl-42, killer cell immunoglobulin-like receptor 2DL1, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1, killer Ig receptor, killer inhibitory receptor 2-2-1, KIR221, MHC class I NK cell receptor, Natural killer-associated transcript 1, NKAT, NKAT-1, NKAT1KIR-K64, p58 killer cell inhibitory receptor KIR-K64, p58 natural killer cell receptor clones CL-42/47.11, p58 NK cell inhibitory receptor NKR-K6,47.11, p58 NK receptor CL-42/47.11, p58.1, p58.1 MHC class-I-specific NK receptor | |
Mouse | |
Protein G purified | |
Immunology, Innate Immunity, Signal Transduction | |
3802.0 | |
Human | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title