Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIR2DL1/CD158a Mouse anti-Human, Allophycocyanin/Cy7, Clone: MM0438-11G25, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP211758APCCY7

Catalog No. N211758APC7


Add to cart

Description

Description

KIR2DL1/CD158a Monoclonal antibody specifically detects KIR2DL1/CD158a in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

KIR2DL1/CD158a
MM0438-11G25
Flow Cytometry
Conjugated, Purified
KIR2DL1
Human KIR
Protein G purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
This antibody detects KIR2DL2, KIR2DL3, KIR2DS2, and KIR2DS4.
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
CD158 antigen-like family member A, CD158a antigen, CD158Ankat1, cl-42, killer cell immunoglobulin-like receptor 2DL1, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 1, killer Ig receptor, killer inhibitory receptor 2-2-1, KIR221, MHC class I NK cell receptor, Natural killer-associated transcript 1, NKAT, NKAT-1, NKAT1KIR-K64, p58 killer cell inhibitory receptor KIR-K64, p58 natural killer cell receptor clones CL-42/47.11, p58 NK cell inhibitory receptor NKR-K6,47.11, p58 NK receptor CL-42/47.11, p58.1, p58.1 MHC class-I-specific NK receptor
Mouse
IgG1
0.1 mL
Primary
3802.0
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit