Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KIR2DL2 Mouse anti-Human, Clone: 190IIC311, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA528120

Catalog No. PIMA528120


Add to cart

Description

Description

Ly49 receptors exhibit allelic specificity for MHC class I Ia molecules and are thought to serve to prevent natural killer (NK) cells from attacking normal cells, while allowing them to attack infected or transformed cells in which class I molecules have been down-regulated. These inhibitory receptors are also expressed on subpopulations of mouse CD8+ T cells.
Specifications

Specifications

KIR2DL2
Monoclonal
1 mg/mL
PBS with 0.09% sodium azide
P43627
KIR2DL2
Recombit human KIR2DL3
Protein G
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Western Blot
190IIC311
Unconjugated
KIR2DL2
Liquid
Mouse
IgG2a
200 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
3803
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit