Learn More
KIR2DL5/CD158f Mouse anti-Human, Janelia Fluor™ 646, Clone: UP-R1, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP262222JF646
Catalog No. NB002787
Add to cart
Description
Description
KIR2DL5/CD158f Monoclonal specifically detects KIR2DL5/CD158f in Human samples. It is validated for Flow Cytometry, Immunocytochemistry.
Specifications
Specifications
KIR2DL5/CD158f
UP-R1
Flow Cytometry, Immunocytochemistry
Purified, Conjugated
KIR2DL5A
Human CD158f-Ig fusion protein
Protein A purified
Store at 4°C in the dark.
Monoclonal
The antibody MEM-59 recognizes neuraminidase-sensitive epitope on CD43 (Leukosialin), a 95-135 kDa type I transmembrane glycoprotein (mucin-type) which is involved in lymphocyte activation. CD43 is expressed by platelets and at high levels on the surface of all leukocytes; it is negative on resting B lymphocytes and erythrocytes.
Flow Cytometry, Immunocytochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
CD158F, CD158F1, killer cell immunoglobulin-like receptor 2DL5A, killer cell immunoglobulin-like receptor KIR2DL5A, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5, killer cell immunoglobulin-like receptor, two domains, long cytoplasmic tail, 5A, killer-cell Ig-like receptor, KIR2DL5, KIR2DL5.1, KIR2DL5.3, KIR2DL5B
Mouse
IgG1 Kappa
0.1 mL
Primary
57292
Human