Learn More
Kir3.1 (KCNJ3) (extracellular) Mouse anti-Human, Mouse, Clone: 3C3, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA527546
Catalog No. PIMA527546
Add to cart
Description
Description
Product is shipped at room temperature as a lyophilized powder and should be stored at -20 °C upon receipt. Reconstitution: add 100 µL of deionized water.
This potassium channel is controlled by G proteins. Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. This receptor plays a crucial role in regulating the heartbeat.
Specifications
Specifications
Kir3.1 (KCNJ3) (extracellular)
Monoclonal
1 mg/mL
PBS with 1% BSA and 0.05% sodium azide; pH 7.4
P48549, P63250
KCNJ3
Mouse
IgG1
100 μg
-20°C
Primary
16519, 3760
Immunocytochemistry, Immunofluorescence, Western Blot
3C3
Unconjugated
KCNJ3
Lyophilized
Kcnj3
Synthetic peptide mapping to the extracellular loop of human Kir 3.1
Antigen affinity chromatography
RUO
Antibody
Monoclonal
Human, Mouse