Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kir3.1 Mouse anti-Human, Janelia Fluor™ 549, Clone: OTI1G3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271065JF549

Catalog No. N271065J549


Add to cart

Description

Description

Kir3.1 Monoclonal antibody specifically detects Kir3.1 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

Kir3.1
OTI1G3
Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
KCNJ3
Recombinant protein fragment corresponding to amino acids 279-501 of human Kir3.1 (NP_002230) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry
Janelia Fluor 549
50mM Sodium Borate with 0.05% Sodium Azide
G protein-activated inward rectifier potassium channel 1, GIRK-1, GIRK1Kir3.1, Inward rectifier K(+) channel Kir3.1, inward rectifier K+ channel KIR3.1, KGA, KIR3.1, Potassium channel, inwardly rectifying subfamily J member 3, potassium inwardly-rectifying channel, subfamily J, member 3
Mouse
IgG1
0.1 mL
Primary
3760.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit