Learn More
KIR3DL3/CD158z Rabbit anti-Human, Alexa Fluor 405, Clone: 1136B, R&D Systems™
Rabbit MonoclonalAntibody
Manufacturer:  R&D Systems FAB8919V100UG
Catalog No. FAB8919V100
Add to cart
This item is not returnable. View return policy
Description
Description
Recombinant Monoclonal Antibody.
Specifications
Specifications
KIR3DL3/CD158z
Monoclonal
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Rabbit
IgG
RUO
Primary
115653
Human
Flow Cytometry
1136B
Flow Cytometry 0.25-1 ug/10^6 cells
Q8N743
Human embryonic kidney cell line HEK293-derived recombinant human KIR3DL3/CD158z Met1-Leu322 Accession # Q8N743
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Detects human KIR3DL3/CD158z in direct ELISAs.
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)