Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human KIR3DL3/CD158z Alexa Fluor 594-conjugated Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Supplier: R&D Systems FAB8919T100UG
This item is not returnable.
View return policy
Description
KIR3DL3/CD158z Monoclonal specifically detects KIR3DL3/CD158z in Human samples. It is validated for Flow Cytometry.Specifications
KIR3DL3/CD158z | |
Monoclonal | |
Alexa Fluor 594 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Rabbit | |
100 μg | |
Primary | |
Detects human KIR3DL3/CD158z in direct ELISAs. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Flow Cytometry | |
1136B | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
Q8N743 | |
Human embryonic kidney cell line HEK293-derived recombinant human KIR3DL3/CD158z Met1-Leu322 Accession # Q8N743 | |
RUO | |
115653 | |
Human | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction