Learn More
Invitrogen™ KIR3DL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA587606
Description
Immunogen sequence: QDKPFLSAWP GTVVSEGQHV TLQCRSRLGF NEFSLSKEDG MPVPELYNRI FRNSFLMGPV TPAHAGTYRC CSSHPHSPTG WSAPSNPVVI MVTGVHRKPS LLAHPGPLVK SGETVILQCW SDVRFERFLL HREGITEDPL RLVGQLHDAG SQVNYSMGPM TPALAGTYRC FGSVTHLPYE LSAPSDPLDI VVVGLYGKPS LSAQPGPTVQ AGENVTLSCS SRSLFDIYHL SREAEAGELR LTAVLRVNGT FQANFPLGPV THGGNYRCFG SFRALPHAWS DPSDPLPVSV TGNSRNL; Positive Samples: HepG2, SGC-7901; Cellular Location: Cell membrane, Single-pass type I membrane protein.
KIR3DL3 is transmembrane glycoproteins expressed by natural killer cells and subsets of T cells. This protein are polymorphic and highly homologous and they are found in a cluster on chromosome 19q13.4 within the 1 Mb leukocyte receptor complex (LRC).
Specifications
KIR3DL3 | |
Polyclonal | |
Unconjugated | |
KIR3DL3 | |
CD158 antigen-like family member Z; CD158Z; killer cell Ig-like receptor KIR3DL7; killer cell immunoglobulin like receptor, three Ig domains and long cytoplasmic tail 3; killer cell immunoglobulin-like receptor 3DL3; killer cell immunoglobulin-like receptor two domains short cytoplasmic tail 2; killer cell immunoglobulin-like receptor, three domains, long cytoplasmic tail, 3; killer cell inhibitory receptor 1; killer-cell immunoglobulin-like receptor 3DL3; KIR2DS2; KIR3DL3; KIR3DL7; KIR44; KIRC1; natural killer cell receptor | |
Rabbit | |
Affinity Chromatography | |
RUO | |
115653 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
1.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N743 | |
KIR3DL3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 26-322 of human KIR3DL3 (NP_7031443). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.