Learn More
Kir6.1 Mouse anti-Human, Mouse, Rat, Gerbil, Alexa Fluor 488, Clone: S366-60, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP259324AF488
Catalog No. NB259324488
Description
Description
Kir6.1 Monoclonal specifically detects Kir6.1 in Human, Mouse, Rat, Gerbil samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Kir6.1
S366-60
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
KCNJ8
Fusion protein amino acids 306-424 (Cytoplasmic C-terminus) of rat Kir6.1
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 45kDa.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Alexa Fluor 488
50mM Sodium Borate with 0.05% Sodium Azide
ATP-sensitive inward rectifier potassium channel 8, Inward rectifier K(+) channel Kir6.1, inwardly rectifying potassium channel KIR6.1, KIR6.1, Potassium channel, inwardly rectifying subfamily J member 8, potassium inwardly-rectifying channel, subfamily J, member 8, uKATP-1
Mouse
IgG2a
100 ul
Primary
3764
Human, Mouse, Rat, Gerbil