Learn More
Kir6.1 Mouse anti-Human, Mouse, Rat, DyLight 405, Clone: S366-60, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259324V
Catalog No. NBP259324V
Add to cart
Description
Description
Kir6.1 Monoclonal antibody specifically detects Kir6.1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
Specifications
Kir6.1
S366-60
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purified
KCNJ8
Fusion protein amino acids 306-424 (Cytoplasmic C-terminus) of rat Kir6.1
Protein G purified
Store at 4C in the dark.
Monoclonal
Detects 45kDa.
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
ATP-sensitive inward rectifier potassium channel 8, Inward rectifier K(+) channel Kir6.1, inwardly rectifying potassium channel KIR6.1, KIR6.1, Potassium channel, inwardly rectifying subfamily J member 8, potassium inwardly-rectifying channel, subfamily J, member 8, uKATP-1
Mouse
IgG2a
100 ul
Primary
3764
Human, Mouse, Rat