Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Kir7.1 (KCNJ13) Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5119868
This item is not returnable.
View return policy
Description
Positive Control: Rat brain tissue lysate, mouse brain tissue lysate, mouse cerebellum tissue lysate, human small intestine tissue. Subcellular Location: Membrane.
Inward rectifier potassium channels are characterized by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. KCNJ13 has a very low single channel conductance, low sensitivity to block by external barium and cesium, and no dependence of its inward rectification properties on the internal blocking particle magnesium.
Specifications
Kir7.1 (KCNJ13) | |
Polyclonal | |
Unconjugated | |
KCNJ13 | |
Inward rectifier K(+) channel Kir7.1; inward rectifier potassium channel 13; Kcnj13; KIR1.4; KIR7.1; LCA16; Potassium channel, inwardly rectifying subfamily J member 13; potassium channel, inwardly rectifying subfamily J, member 13; potassium inwardly-rectifying channel, subfamily J, member 13; potassium voltage-gated channel subfamily J member 13; SVD | |
Rabbit | |
Affinity chromatography | |
RUO | |
100040591, 3769, 94341 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
O60928, O70617, P86046 | |
KCNJ13 | |
Recombinant protein within C-terminal human KCNJ13/Kir7.1 from aa 200-aa360. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction