Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kirrel1/NEPH1 Rabbit anti-Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen Kirrel1/NEPH1
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094756
View Documents
Novus Biologicals
NBP303509100UL
100 μg
Each for $320.71
Add to cart
 
NB094757
View Documents
Novus Biologicals
NBP30350920UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

Kirrel1/NEPH1 Polyclonal antibody specifically detects Kirrel1/NEPH1 in Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

Kirrel1/NEPH1
Unconjugated
Rabbit
Mouse, Rat
FLJ10845, kin of IRRE like (Drosophila), MGC129542, MGC129543, NEPH1, nephrin-like protein 1
Recombinant fusion protein containing a sequence corresponding to amino acids 17-200 of human Kirrel1/NEPH1 (NP_060710.3).
Polyclonal
Affinity purified
Western Blot
Purified
RUO
PBS with 50% glycerol, pH7.3.
55243
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit