Learn More
Aviva Systems Biology Corp KIRREL2 Antibody - N-terminal region (ARP50035_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50035_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KIRREL2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KIRREL2. Alternate names: DKFZP564A1164, FILTRIN, MGC15718, NEPH3, NLG1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60248