Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIRREL2 Monoclonal Antibody (318311)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA524040
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a type I transmembrane protein and member of the immunoglobulin superfamily of cell adhesion molecules. The encoded protein localizes to adherens junctions in pancreatic beta cells and regulates insulin secretion. Autoantibodies against the encoded protein have been detected in serum from patients with type 1 diabetes. This gene may also play a role in glomerular development and decreased expression of this gene has been observed in human glomerular diseases. This gene and the related opposite-strand gene nephrin are regulated by a bidirectional promoter.
Specifications
KIRREL2 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q6UWL6, Q7TSU7 | |
KIRREL2 | |
Mouse myeloma cell line NS0-derived recombinant human Kirrel2 Gly21-Arg503. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |
Western Blot | |
318311 | |
Unconjugated | |
KIRREL2 | |
C330019F22Rik; DKFZp564A1164; FILTRIN; kin of IRRE like 2; kin of IRRE like 2 (Drosophila); kin of irregular chiasm-like 2; kin of irregular chiasm-like protein 2; kin of IRRE-like 2; kin of IRRE-like protein 2; kin of IRRE-like protein 2; LOW QUALITY PROTEIN: kin of IRRE-like protein 2; kirre like nephrin family adhesion molecule 2; KIRREL2; MGC15718; NEPH3; nephrin-like 3; nephrin-like gene 1; nephrin-like protein 3; NLG1; UNQ5827/PRO19646; X kin of IRRE like 2 | |
Mouse | |
Protein A/G | |
RUO | |
243911, 84063 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction