Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KISS1 Polyclonal Antibody
KISS1 Polyclonal Antibody
Supplier: Thermo Scientific PA5106920
Description
KISS1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

Specifications
KISS1 | |
Polyclonal | |
Unconjugated | |
KISS1 | |
0610012C01Rik; Eseptin; HH13; KISS1; KiSS-1; KiSS-1 metastasis-suppressor; Kiss1 metastasis-suppressor; Kiss1 variant E1a-E2b-E3; Kiss1 variant E1b-E2a-E3; Kiss1 variant E1b-E2b-E3; kisspeptin; Kisspeptin-1; Kisspeptin-10; Kisspeptin-13; Kisspeptin-14; Kisspeptin-52; Kisspeptin-54; malignant melanoma metastasis-suppressor; metastasis-suppressor KiSS-1; metastatin; metastin; Metastin45-54; MLL septin-like fusion; PP5098; prepro-kisspeptin | |
Rabbit | |
Affinity Chromatography | |
RUO | |
280287, 289023, 3814 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q15726, Q6Y4S4, Q7TSB7 | |
KISS1 | |
A synthesized peptide derived from human KISS1(Accession Q15726), corresponding to amino acid residues A88-G138. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction