Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ KiSS1R/GPR54 Recombinant Protein Antigen

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

Manufacturer:  Novus Biologicals™ NBP257942PEP

Catalog No. NBP257942PE


Add to cart

Description

Description

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human KiSS1R/GPR54. Source: E.coli Amino Acid Sequence: VATSGPNASWGAPANASGCPGCGANASDGPVPSPRAVDAWLV The KiSS1R/GPR54 Recombinant Protein Antigen is derived from E. coli. The KiSS1R/GPR54 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications

Specifications

Blocking/Neutralizing, Control
84634
>80% by SDS-PAGE and Coomassie blue staining
E.coli
RUO
KISS1R
Unlabeled
Yes
PBS and 1M Urea, pH 7.4
KiSS1R/GPR54 Recombinant Protein Antigen
100μL
Store at −20°C. Avoid freeze-thaw cycles
AXOR12hypogonadotropin-1, G protein-coupled receptor 54, GPR54kiSS-1R, G-protein coupled receptor 54, G-protein coupled receptor OT7T175, HOT7T175, Hypogonadotropin-1, kiSS-1 receptor, KISS1 receptor, KiSS-1R, Kisspeptins receptor, Metastin receptor
Recombinant Protein Antigen
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For research use only.