Learn More
Alfa Aesar™ Kisspeptin-10
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J65487EXD
 View more versions of this product
Catalog No. AAJ65487EXD
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
1302.44
0.5mg
Tyr-Asn-Trp-Asn-Ser-Phe-Gly-Leu-Arg-Phe-NH2
-30° to -10°C
Kisspeptin-10
HPLC
Powder
RUO
Metastin (45-54), amide, human