Learn More
DWK Life Sciences MicroLiter 9mm Screwthread Autosampler Vial, Cap and Septa Unassembled Component Kits with FEP/Natural Rubber Septa
Unassembled component kits offer the convenience of one product number for the vial, cap, and septa. Components are packaged to work with the unique MicroLiter Dispenser.
Manufacturer:  DWK Life Sciences 091220Y
 View more versions of this product
Catalog No. 15-111-257
Add to cart
Description
Description
Available in multiple colors and septa formats and with or without writing patch.
Specifications
Specifications
Yellow
Borosilicate Glass (Type 1)
2 mL
FEP/Natural Rubber/FEP
Vial Kit
Screw Thread
9 mm
12 mm
No
FEP/Natural Rubber
100
Polypropylene
32 mm