Learn More
Kit Oil no. 11 for Oil Repellency on Paper, Reagents
Kit Oil no. 11 for Oil Repellency on Paper, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier:  Reagents Holdings Llc CK1972001C
Quantity 1 L
Catalog No. 01-334-860
Only null left
Add to Cart
Description
Description
Specifications
Chemical Identifiers
108-88-3 , 142-82-5
92.14
toluene
C7H8
YXFVVABEGXRONW-UHFFFAOYSA-N
CC1=CC=CC=C1
Specifications
Colorless
C7H8
CC1=CC=CC=C1
92.14
Glass Bottle
1 L
Liquid
108-88-3 , 142-82-5
YXFVVABEGXRONW-UHFFFAOYSA-N
toluene
Reagent
0.7728
Non-sterile
Kit Oil #11 for Oil Repellency on Paper
Product Suggestions
Product Suggestions