Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ KIZUNA Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5106259
Description
Antibody detects endogenous levels of total Kizuna.
Centrosomal protein required for establishing a robust mitotic centrosome architecture that can endure the forces that converge on the centrosomes during spindle formation. Required for stabilizing the expanded pericentriolar material around the centriole.
Specifications
KIZUNA | |
Polyclonal | |
Unconjugated | |
KIZ | |
C20orf19; Centrosomal protein kizuna; Gm114; HT013; KIZ; Kizuna; kizuna centrosomal protein; NCRNA00153; PLK1S1; polo-like kinase 1 substrate 1; RGD1307071; RP69 | |
Rabbit | |
Affinity chromatography | |
RUO | |
228730, 55857 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q2M2Z5, Q3UXL4 | |
KIZ | |
A synthesized peptide derived from human KIZ(Accession Q2M2Z5), corresponding to amino acid residues S357-G407. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction