Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ KJ Pyr 9
High affinity Myc inhibitor
$179.80 - $747.04
Specifications
Chemical Name or Material | 4-[2-(2-Furanyl)-6-(4-nitrophenyl)-4-pyridinyl]benzamide |
---|---|
CAS | 581073-80-5 |
Target | Additional Transcription Factor Compounds |
Molecular Formula | C22H15N3O4 |
Formula Weight | 385.37 |
Description
High affinity Myc inhibitor (Kd = 6.5 nM). Disrupts Myc-Max interaction. Inhibits Myc-dependent cancer cell line proliferation in vitro. Inhibits growth of MDA-MB-231 breast cancer cell xenografts in mice. Brain penetrant.Specifications
4-[2-(2-Furanyl)-6-(4-nitrophenyl)-4-pyridinyl]benzamide | |
Additional Transcription Factor Compounds | |
385.37 |
581073-80-5 | |
C22H15N3O4 | |
>98% |
Safety and Handling
Recommended Storage : Store at 4°C
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title