Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kjeldahl Solution (Dilute) 25% Sodium Hydroxide/1.25% Sodium Thiosulfate, Reagents

Catalog No. p-200022037
Click to view available options
Quantity:
4 L
10 L
20 L

Kjeldahl Solution (Dilute) 25% Sodium Hydroxide/1.25% Sodium Thiosulfate, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Kjeldahl Solution (Dilute) 25% Sodium Hydroxide/1.25% Sodium Thiosulfate
Color Colorless
Quantity 10 L
CAS 7732-18-5, 1310-73-2
Boiling Point 106°C
Packaging Cubitainer
Physical Form Liquid
Synonym Nitrogen
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.