Learn More
Kjeldahl tablets with selenium (Wieninger), Thermo Scientific Chemicals
$260.00 - $460.00
Specifications
Chemical Name or Material | Kjeldahl tablets with selenium (Wieninger) |
---|---|
CAS | 7782-49-2, 7758-99-8, 7757-82-6 |
MDL Number | MFCD00147338 |
Packaging | Plastic bottle |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
AC243920010
|
Thermo Scientific Chemicals
243920010 |
1 kg | Plastic bottle |
Each for $260.00
Order in Bulk
Request bulk or custom formats |
|
||||
AC243920025
|
Thermo Scientific Chemicals
243920025 |
2.5 kg | Plastic bottle |
Each for $460.00
Order in Bulk
Request bulk or custom formats |
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Kjeldahl tablets with selenium (Wieninger) | |
7782-49-2, 7758-99-8, 7757-82-6 | |
MFCD00147338 | |
Plastic bottle |
Safety and Handling
GHS H Statement
Harmful to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
EINECSNumber : 231-957-4
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.