Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kjeltabs S-5, Foss™

Click to view available options
Quantity:
1000 tablets
Packaging:
5g
7782-49-2
Se
78.971
BUGBHKTXTAQXES-UHFFFAOYSA-N
6326970
CHEBI:27568
selenium
[Se]
7782-49-2
Se
78.971
BUGBHKTXTAQXES-UHFFFAOYSA-N
6326970
CHEBI:27568
selenium
[Se]

Composition per Tablet: 5g K2SO4 + 5mg Se

Quantity 1000 tablets
Packaging 5g
Physical Form Flat Round Speckled Tablets
Chemical Name or Material Selenium Kjeltabs