Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KL 001, Tocris Bioscience™

Catalog No. p-4816819
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Cryptochrome protein stabilizer; lengthens circadian period

Cryptochrome protein (CRY) stabilizer; blocks proteasome-mediated degradation of CRY1 and CRY2. Lengthens circadian period in mouse NIH-3T3 fibroblasts without altering casein kinase activity. Represses glucagon-induced hepatic gluconeogenesis through the induction of Pck1 and G6pc expression.

Chemical Identifiers

CAS 309928-48-1
Molecular Formula C21H22N2O4S
Molecular Weight (g/mol) 398.477
InChI Key OQAFDLPAPSSOHY-UHFFFAOYSA-N
Synonym n-3-9h-carbazol-9-yl-2-hydroxypropyl-n-furan-2-ylmethyl methanesulfonamide, n-3-carbazol-9-yl-2-hydroxypropyl-n-furan-2-ylmethyl methanesulfonamide, cbmicro_005790, 10.14272/oqafdlpapssohy-uhfffaoysa-n, doi:10.14272/oqafdlpapssohy-uhfffaoysa-n, 3-carbazol-9-yl-2-hydroxypropyl 2-furylmethyl methylsulfonyl amine, n-3-carbazol-9-yl-2-hydroxy-propyl-n-2-furfuryl methanesulfonamide, n-3-carbazol-9-yl-2-hydroxy-propyl-n-furan-2-ylmethyl-methanesulfonamide
PubChem CID 2888648
IUPAC Name N-(3-carbazol-9-yl-2-hydroxypropyl)-N-(furan-2-ylmethyl)methanesulfonamide
SMILES CS(=O)(=O)N(CC1=CC=CO1)CC(CN2C3=CC=CC=C3C4=CC=CC=C42)O

Specifications

Quantity 10 mg
Formula Weight 398.48
Percent Purity >98%
Chemical Name or Material KL 001
Recommended Storage Store at +4°C
Provide Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.