Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ KLB Polyclonal Antibody

Rabbit Polyclonal Antibody

$209.50 - $511.50

Specifications

Antigen KLB
Concentration 0.5 mg/mL
Content And Storage -20°C
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PIPA5119246
View Documents
Invitrogen™
PA5119246
200 μL
Each for $511.50
Only null left
Add to Cart
 
PIPA5121806
View Documents
Invitrogen™
PA5121806
20 μL
Each for $209.50
Only null left
Add to Cart
 
Description

Description

KLB Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

KLB is a single-pass type III membrane protein. It contributes to the transcriptional repression of cholesterol 7-alpha-hydroxylase (CYP7A1), the rate-limiting enzyme in bile acid synthesis. KLB is probably inactive as a glycosidase. It increases the ability of FGFR1 and FGFR4 to bind FGF21.
Specifications

Specifications

KLB
-20°C
Polyclonal
Liquid
IgG
Human
KLB
AV071179; Betakl; betaKlotho; beta-klotho; BKL; Klb; Klb-ps1; klotho beta; klotho beta like; klotho beta, pseudogene 1; klotho beta-like protein; MGC142213; RGD1308227
KLB
Primary
Antigen affinity chromatography
0.5 mg/mL
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Unconjugated
Rabbit
RUO
PBS with 50% glycerol and 0.05% ProClin 300
Q86Z14
152831
Recombinant protein Klotho Beta. The antigen corresponds to amino acid range 517-636 of the target protein.
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.