Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLC3 Rabbit anti-Human, Polyclonal , Abnova™

Rabbit polyclonal antibody raised against recombinant human KLC3.

Manufacturer:  Abnova Corporation PAB29466

Catalog No. 89-934-407


Add to cart

Description

Description

This gene encodes a member of the kinesin light chain gene family. Kinesins are molecular motors involved in the transport of cargo along microtubules, and are composed of two kinesin heavy chain (KHC) and two kinesin light chain (KLC) molecules. KLCs are thought to typically be involved in binding cargo and regulating kinesin activity. In the rat, a protein similar to this gene product is expressed in post-meiotic spermatids, where it associates with structural components of sperm tails and mitochondria. [provided by RefSeq]

Sequence: MSVQVAAPGSAGLGPERLSPEELVRQTRQVV
Specifications

Specifications

KLC3
Unconjugated
Immunohistochemistry (1:500-1:1000) Immunofluorescence (1-4 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
KLC3
Recombinant protein corresponding to human KLC3.
Antigen affinity purification
RUO
Primary
147700
Immunofluorescence, Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against recombinant human KLC3.
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide)
KLC2/KLC2L/KLCt/KNS2B
Rabbit
IgG
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit