Learn More
Sklar InstrumentsSupplier Diversity Partner Kleegman Dilator, Sklar, Narrow, 10-1/2 in., Curved, Stainless Steel. EA/1. DFS Item
Manufacturer:  Sklar InstrumentsSupplier Diversity Partner 904902
Kleegman Dilator, Sklar, Narrow, 10-1/2 in., Curved, Stainless Steel. Sklar's Kleegman Dilator is a narrow, double-ended OB/GYN instrument used to measure dilation or further dilate the uterus. The tips are smooth, rounded, and curve slightly in opposite directions. The smaller end tapers from 3mm to a 2mm tip. The large end tapers from 3mm to a 2.7mm tip.
Catalog No. 50-143-6750