Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ A30 Breathable Splash Particle Protection Coveralls
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
6X-Large
Description
MICROFORCE™ Barrier SMS Fabric incorporates a web of microfibers that filter particulates and water-based liquids. Suitable for many applications including pharmaceutical research, food processing and more.
- Excellent protection from nonhazardous particles and liquids
- Strong and abrasion resistant; suitable for multiple wearings
- Seamless front to provide more protection in the primary exposure areas
- Zipper front with 1 in. (2.5cm) flap and elastic back
- Color: White
- With elastic back, wrists and ankles
Compliance
Meets NFPA™ 99 Health Care Facilities criteria for antistatic materials.
Specifications
Size | 6X-Large |
Material | Polypropylene |
Color | White |
Ankle Style | Elastic |
Cuff Style | Elastic |
Back Style | Elastic |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction