Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ A40 Reflex™ Coveralls
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
Description
KleenGuard™ A40 Reflex Coveralls™ with Hood comes with a respirator fit hood, zipper front, and elastic wrists and ankles. Made of breathable, microporous film to keep out debris, non-hazardous liquid chemicals and dry particulates.
These coveralls with hood pass NFPA 99 for antistatic materials, pass ASTM F1670 testing for penetration of blood and bodily fluids, and have Reflex sizing for a more comfortable fit and less rip outs. Popular industries using this product include aviation, pharmaceutical manufacturing, liquid handling, general manufacturing and so much more.
- 25 Garments/Case
- KleenGuard™ A40 Reflex™ Coveralls are made of microporous film laminate with a respirator fit hood, zipper front and elastic wrists and ankles
- These hooded coveralls offer better liquid and particulate barrier than the competition
- Made to keep out debris, dry particulates and liquid splashes
- A40 KleenGuard™ protective suits pass NFPA 99 criteria for antistatic materials
Specifications
Size | Large |
Gender | Unisex |
Material | Microporous Film Laminate |
Color | White |
Disposable | Yes |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Closure Type | Zipper |
Seam Type | Serged |
Show More |