Learn More
Kimberly-Clark Professional™ KleenGuard™ A60 Ultra Bloodborne Pathogen and Chemical Splash Protection Hoods Available on GSA/VA Contract for Federal Government customers only.
KleenGuard™ A60 Bloodborne Pathogen and Chemical Splash Protection Hoods are designed for sensitive jobs like working in research, crime labs, crime scene cleanup, forensics, embalming, and emergency medical response
Manufacturer:  Kimberly-Clark Professional™ 45343
Catalog No. 19-057-445
Add to cart
Description
Description
  • Passes ASTM F1670/F1671 testing for penetration of blood, body fluids and bloodborne pathogens
  • Elastic seal around the face opening
  • Serged seams
  • Three-layer fabric construction features a middle layer of microporous film that allows heat and sweat vapor to escape, keeping the worker cool and outer layer film barrier against tearing and abrasion
  • Passes NFPA 99 for antistatic materials
  • Recyclable via The RightCycle Program by Kimberly-Clark Professional
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Passes ASTM F1670/F1671 testing for penetration of blood, body fluids and bloodborne pathogens, Passes NFPA 99 for antistatic materials
Blue
100/Cs.
Hood
Safety
Yes
Animal research; Emergency medical response; Crime scene cleanup; Embalming/Forensics; Tank Cleaning; Fiberglass Manufacturing; Liquid Handling; Food Processing
One size fits all