Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ A80 Chemical Permeation and Jet Liquid Protection Coveralls
For chemical permeation and jet liquid protection
Supplier: Kimberly-Clark Professional™ 45643
This item is not returnable.
View return policy
Description
Level B/C suit made from durable fabric with exterior laminated with Saranex™ 23-P film. Suitable for numerous applications including chemical handling/mixing/clean-up, hazardous material handling, hazardous waste cleanup, industrial Hazmat teams, petrochemical, and utilities.
- Zipper front with taped storm flap
- Interior has comfortable, cloth-like feel against the skin
- Reflex™ Design provides 12% more chest room and 6% longer body length
Specifications
Large | |
White | |
Elastic | |
Attached | |
Taped | |
Hooded (respirator-fit), zipper front with storm flap, elastic wrist and ankle, taped storm flap, Reflex™ design; Saranex™ 23-P Film Laminated to Durable Fabric | |
1.15 lb. |
Saranex™ 23-P fabric | |
Elastic | |
Hooded | |
Front zipper with storm flap | |
Chemical | |
Chemical handling or mixing or cleanup, hazardous material handling, hazardous waste cleanup, industrial hazmat teams, petrochemicals, utilities and for chemical permeation and jet liquid protection | |
0.52 kg |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title