Learn More
Kimberly-Clark Professional™ KleenGuard™ A80 Chemical Permeation and Jet Liquid Protection Coveralls
For chemical permeation and jet liquid protection
Supplier:  Kimberly-Clark Professional™ 45663
 View more versions of this product
Catalog No. 19-057-450A
Add to cart
This item is not returnable. View return policy
Description
Description
Level B/C suit made from durable fabric with exterior laminated with Saranex™ 23-P film. Suitable for numerous applications including chemical handling/mixing/clean-up, hazardous material handling, hazardous waste cleanup, industrial Hazmat teams, petrochemical, and utilities.
  • Zipper front with taped storm flap
  • Interior has comfortable, cloth-like feel against the skin
  • Reflex™ Design provides 12% more chest room and 6% longer body length
Specifications
Specifications
Large
White
Elastic
Attached
Taped
Hooded (respirator-fit), zipper front with storm flap, elastic wrist and ankle, exterior lamination; Saranex™ 23-P Film Laminated to Durable Fabric
1.33 lb.
Saranex™ 23-P fabric
Attached boots
Hooded
Front zipper with storm flap
Chemical
Chemical handling or mixing or cleanup, hazardous material handling, hazardous waste cleanup, industrial hazmat teams, petrochemicals, utilities and for chemical permeation and jet liquid protection
0.6 kg