Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Kimberly-Clark Professional™ KleenGuard™ A80 Chemical Permeation and Jet Liquid Protection Coveralls
Click to view available options
Size:
Large
X-Large
2X-Large
3X-Large
4X-Large
Hood Type:
Attached
Elastic
Ankle Style:
Attached Boots
Elastic
Seam Type:
Taped
Description
Level B/C suit made from durable fabric with exterior laminated with Saranex™ 23-P film. Suitable for numerous applications including chemical handling/mixing/clean-up, hazardous material handling, hazardous waste cleanup, industrial Hazmat teams, petrochemical, and utilities.
- Zipper front with taped storm flap
- Interior has comfortable, cloth-like feel against the skin
- Reflex™ Design provides 12% more chest room and 6% longer body length
Specifications
Size | 4X-Large |
Material | Saranex 23-P Fabric |
Color | White |
Ankle Style | Attached Boots |
Cuff Style | Elastic |
Neck Style | Hooded |
Hood Type | Attached |
Closure Type | Front Zipper with Storm Flap |
Seam Type | Taped |
Resistance | Chemicals |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction