Learn More
Kimberly-Clark Professional™ KleenGuard™ Calico Safety Glasses
Click to view available options
Lens Tint:
Amber
Smoke
Description
- Wraparound design and impact-resistant lenses
- Interchangeable head strap and temples allow customizatio for a closer fit or enhanced comfort
- Lenses feature EN166 N anti-fog coating and fog shield
- Cushioned brow bar for additional comfort and ventilation
- Lens options include:
- Amber anti-fog lenses with light gathering properties ideal for low-light and low-contrast environments
- Smoke anti-fog lenses ideal for bright, sunny days to reduce glare
Specifications
Certifications/Compliance | EN166 1FT, CSA, ANSI Z87.1+ |
Lens Coating | Impact-resistant, UV-treated |
Lens Tint | Smoke |
For Use With (Application) | Areas where there's a high risk for airborne debris |
Frame Color | Black |
No. per Case | 12 pairs/Cs. |
Lens Material | Polycarbonate |
Frame Material | Plastic with foam brow bar |
UV Protection | 0.999 |