Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORS Nasco Klein™ Wire Strippers

Make clean cuts with little effort

Supplier:  ORS Nasco 4091011

Catalog No. 19-313-570


Add to cart

Description

Description

  • Serrated nose jaws to bend, shape, or pull wire
  • Holes for looping wire ends
  • Coil-spring opening action reduces fatigue
  • Black oxide finish, blue handle
  • Cutter size: 0.5 in.
  • Strips solid wire: 10 AWG to 20 AWG
  • Length: 6 in.

Specifications

Specifications

0.5 in.
6 in.
15.24 cm
Coil spring opening action, Plastic covered blue handles
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit