Learn More
Thermo Scientific™ Klenow Fragment, exo- Promotion
Klenow Fragment, exo- is the large fragment of DNA polymerase I. Exhibits 5'3' polymerase activity, but lacks the 3'5' and 5'3' exonuclease activities.
Manufacturer:  Thermo Scientific™ EP0421
 View more versions of this product
Catalog No. FEREP0421PM
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Klenow Fragment, exo-, is the large fragment of DNA polymerase I . It exhibits 5'3' polymerase activity, but lacks the 3'5' and 5'3' exonuclease activities of DNA Polymerase I. The 3'5' exonuclease activity of the enzyme is eliminated by mutations in the 3'5'-exonuclease active sit.
Highlights
  • Lacks 3'5' exonuclease activity
  • Incorporates modified nucleotides (e.g., Cy3-, Cy5-, fluorescein-, rhodamine-, aminoallyl-, biotin-labeled nucleotides)
  • Active in restriction enzyme, PCR, and RT buffers
Specifications
Specifications
5U/μL
E.coli cells with a cloned DNA fragment of the mutated polA gene
300U
metal chelators, PPi, Pi (at high concentrations) (7)
68kDa monomer