Learn More
Thermo Scientific™ Klenow Fragment Promotion
Klenow Fragment is the Large Fragment of DNA Polymerase exhibiting 5'3' polymerase, 3'5' exonuclease (proofreading) activities, but lacking 5'3' exonuclease activity.
Manufacturer:  Thermo Scientific™ EP0051
 View more versions of this product
Catalog No. FEREP0051PM
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Klenow Fragment is the large fragment of DNA polymerase I. It exhibits 5'3' polymerase activity and 3'5' exonuclease (proofreading) activity, but lacks 5'3' exonuclease activity of DNA polymerase I.
Highlights
  • Incorporates modified nucleotides (e.g., Cy3-, Cy5-, aminoallyl-, biotin-, digoxigenin- and fluorescently-labeled nucleotides)
  • Active in restriction enzyme, PCR, RT, and T4 DNA Ligase buffers
Specifications
Specifications
10U/μL
E.coli cells with a cloned fragment of the polA gene
300U
metal chelators, PPi, Pi (at high concentrations) (8)
68kDa monomer