Learn More
KLF1 Mouse anti-Human, Alexa Fluor 700, Clone: 1B6A3, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237380AF700
Catalog No. NBP237380X
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
KLF1
1B6A3
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
Purified
KLF1
Purified recombinant fragment of human KLF1 (AA: 208-362) expressed in E. Coli.
Protein A or G purified
Cancer
Primary
10661
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
EKLFerythroid-specific transcription factor EKLF, Erythroid krueppel-like transcription factor, erythroid Kruppel-like factor, HBFQTL6, INLU, Krueppel-like factor 1, Kruppel-like factor 1 (erythroid), monoclonal A3D8
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human