Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF1 Mouse anti-Human, Biotin, Clone: 1B6A3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237380B

Catalog No. NBP237380B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

KLF1
1B6A3
Western Blot, ELISA, Immunohistochemistry
Purified
KLF1
Purified recombinant fragment of human KLF1 (AA: 208-362) expressed in E. Coli.
Protein A or G purified
Cancer
Primary
10661
ELISA, Immunohistochemistry, Western Blot
Biotin
PBS with 0.05% Sodium Azide
EKLFerythroid-specific transcription factor EKLF, Erythroid krueppel-like transcription factor, erythroid Kruppel-like factor, HBFQTL6, INLU, Krueppel-like factor 1, Kruppel-like factor 1 (erythroid), monoclonal A3D8
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit