Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF11 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP31044725UL

 View more versions of this product

Catalog No. NB125796


Add to cart

Description

Description

KLF11 Polyclonal specifically detects KLF11 in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

KLF11
Unconjugated
PBS buffer, 2% sucrose
FKLFFKLF1, Krueppel-like factor 11, Kruppel-like factor 11, MODY7TGFB-inducible early growth response protein 2, TIEG-2, TIEG2TGFB inducible early growth response 2, Tieg3, Transforming growth factor-beta-inducible early growth response protein 2
The immunogen is a synthetic peptide corresponding to a region of Mouse (NP_848134). Peptide sequence PASGSSCRAVMTSVIRHTGESPAPTRFPTGPTQEQRASDSGEGQERLLDH
25 μg
Cell Cycle and Replication, Transcription Factors and Regulators
Primary
8462
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Affinity purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Mouse
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit