Learn More
KLF14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 212341AP
Catalog No. 50-173-3331
Add to cart
Description
Description
SP6 belongs to a family of transcription factors that contain 3 classical zinc finger DNA-binding domains consisting of a zinc atom tetrahedrally coordinated by 2 cysteines and 2 histidines (C2H2 motif). These transcription factors bind to GC-rich sequences and related GT and CACCC boxes.SP6 belongs to a family of transcription factors that contain 3 classical zinc finger DNA-binding domains consisting of a zinc atom tetrahedrally coordinated by 2 cysteines and 2 histidines (C2H2 motif). These transcription factors bind to GC-rich sequences and related GT and CACCC boxes (Scohy et al., 2000).
Specifications
Specifications
KLF14
0.22 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q3SY56, Q9ESX2
EPFN, EPIPROFIN, KLF14, Krueppel like factor 14, SP6, Sp6 transcription factor, Transcription factor Sp6
Rabbit
IgG
150 μL
-20°C
Primary
363672, 80320, 83395
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
SP6
Liquid
Sp6
KLF14 Fusion Protein Ag15707
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat