Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF16 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$106.19 - $352.89

Specifications

Antigen KLF16
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB126672
View Documents
Novus Biologicals
NBP31088525UL
25 μg
Each for $106.19
Add to cart
 
NB126671
View Documents
Novus Biologicals
NBP310885100UL
100 μg
Each for $352.89
Add to cart
 
Description

Description

KLF16 Polyclonal specifically detects KLF16 in Human samples. It is validated for Western Blot.
Specifications

Specifications

KLF16
Western Blot
Purified
RUO
PBS buffer, 2% sucrose
83855
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Human
basic transcription element binding protein 4, Basic transcription element-binding protein 4, BTEB4Transcription factor NSLP2, dopamine receptor regulating factor, DRRF, Krueppel-like factor 16, Kruppel-like factor 16, Novel Sp1-like zinc finger transcription factor 2, NSLP2BTE-binding protein 4, Transcription factor BTEB4
The immunogen is a synthetic peptide directed towards the middle region of human KLF16 (NP_114124). Peptide sequence GAAAAPHLLAASILADLRGGPGAAPGGASPASSSSAASSPSSGRAPGAAP
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit