Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KLF17 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA550340

Catalog No. PA550340


Add to cart

Description

Description

The antibody detects endogenous levels of total KLF17 protein.

Transcription repressor that binds to the promoter of target genes and prevents their expression. Acts as a negative regulator of epithelial-mesenchymal transition and metastasis in breast cancer. Specifically binds the 5 ft.-CACCC-3 ft. sequence in the promoter of ID1, a key metastasis regulator in breast cancer, and repress its expression. May be a germ cell-specific transcription factor that plays important roles in spermatid differentiation and oocyte development.
Specifications

Specifications

KLF17
Polyclonal
Unconjugated
KLF17
Liquid
KLF17
Fusion protein corresponding to residues near the C terminal of human Kruppel-like factor 17
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunohistochemistry (Paraffin)
1.3 mg/mL
PBS with 50% glycerol and 0.05% sodium azide; pH 7.3
Q5JT82
ZNF393
Rabbit
IgG
100 μL
-20°C
Primary
128209
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit