Learn More
KLF2 Mouse anti-Human, Clone: OTI6A3, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF806991
Catalog No. 50-166-8066
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
KLF2 belongs to the krueppel C2H2-type zinc-finger protein family. It contains 3 C2H2-type zinc fingers. KLF2 binds to the CACCC box in the beta-globin gene promoter and activates transcription.
Specifications
Specifications
KLF2
Monoclonal
Unconjugated
KLF2
Lyophilized
KLF2
Full length human recombit protein of human KLF2 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Immunohistochemistry (Paraffin), Western Blot
OTI6A3
PBS with 8% trehalose and no preservative; pH 7.3
Q9Y5W3
LKLF
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
10365