Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
KLF2 Mouse anti-Human, Mouse, Clone: 2F12A11, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP261812
Description
KLF2 Monoclonal antibody specifically detects KLF2 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry.Specifications
KLF2 | |
2F12A11 | |
Unconjugated | |
Purified | |
Krueppel-like factor 2, Kruppel-like factor 2 (lung), Kruppel-like factor LKLF, LKLFLung krueppel-like factor, lung Kruppel-like zinc finger transcription factor | |
KLF2 | |
Purified recombinant fragment of human KLF2 (AA: 251-355) expressed in E. Coli. | |
37.4 kDa | |
Primary | |
0.1 ml | |
Human,Mouse |
Western Blot,Flow Cytometry,ELISA,Immunohistochemistry | |
1 mg/mL | |
Western Blot 1:100-1:500, Flow Cytometry, ELISA 1:10000 | |
PBS with 0.05% Sodium Azide | |
10365 | |
Mouse | |
IgG1 | |
Monoclonal | |
Protein G purified | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title